![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | PPP1R1C |
Antigen Name | Protein Phosphatase 1, Regulatory Subunit 1C |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Mouse |
Western Blot | Yes |
Antigen Synonyms | IPP5 |
Gene ID | 151242 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |