Anti-ZNF385B Mouse Polyclonal Antibody

H00151126-B01P
10574-680EA 316.92 USD
10574-680
Anti-ZNF385B Mouse Polyclonal Antibody
Type: Primary
Antigen: ZNF385B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR