Anti-COMMD1 Mouse Monoclonal Antibody [clone: 2A12]

Supplier: Abnova
H00150684-M01
10594-062EA 335 USD
10594-062
Anti-COMMD1 Mouse Monoclonal Antibody [clone: 2A12]
Antibodies
Type: Primary
Antigen: COMMD1
Clonality: Monoclonal
Clone: 2A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR