Anti-CCT8L2 Mouse Monoclonal Antibody [clone: 2C9]

Supplier: Abnova
H00150160-M12
10582-004EA 335 USD
10582-004
Anti-CCT8L2 Mouse Monoclonal Antibody [clone: 2C9]
Antibodies
Type: Primary
Antigen: CCT8L2
Clonality: Monoclonal
Clone: 2C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR