![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | SIK1 |
Antigen Name | Salt-inducible Kinase 1 |
Reactivity | Human, Mouse |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Western Blot | Yes |
Antigen Synonyms | SIK-1, Serine/threonine-protein kinase SNF1-like kinase 1, Serine/threonine-protein kinase SNF1LK, Serine/threonine-protein kinase SIK1, SNF1LK, SIK1, Salt-inducible kinase 1, SIK |
Gene ID | 150094 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |