Anti-WFDC5 Mouse Monoclonal Antibody [clone: 3E5]

Supplier: Abnova
H00149708-M08
10605-798EA 335 USD
10605-798
Anti-WFDC5 Mouse Monoclonal Antibody [clone: 3E5]
Antibodies
Type: Primary
Antigen: WFDC5
Clonality: Monoclonal
Clone: 300000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR