Anti-IL23R Mouse Monoclonal Antibody [clone: 1C12]

Supplier: Abnova
H00149233-M02
10590-284EA 335 USD
10590-284
Anti-IL23R Mouse Monoclonal Antibody [clone: 1C12]
Antibodies
Type: Primary
Antigen: IL23R
Clonality: Monoclonal
Clone: 1C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR