Anti-MOB3C Mouse Polyclonal Antibody

H00148932-B01P
10564-526EA 316.92 USD
10564-526
Anti-MOB3C Mouse Polyclonal Antibody
Type: Primary
Antigen: MOB3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR