![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | HFE2 |
Antigen Name | Hemochromatosis Type 2 |
Reactivity | Human, Mouse |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Western Blot | Yes |
Antigen Synonyms | HFE2A, HJV, RGMC, JH |
Gene ID | 148738 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |