Anti-UBE2U Mouse Monoclonal Antibody [clone: 3D4]

Supplier: Abnova
H00148581-M07
10605-256EA 335 USD
10605-256
Anti-UBE2U Mouse Monoclonal Antibody [clone: 3D4]
Antibodies
Type: Primary
Antigen: UBE2U
Clonality: Monoclonal
Clone: 3D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR