Anti-HIPK4 Mouse Monoclonal Antibody [clone: 2E8]

Supplier: Abnova
H00147746-M01
10588-842EA 335 USD
10588-842
Anti-HIPK4 Mouse Monoclonal Antibody [clone: 2E8]
Antibodies
Type: Primary
Antigen: HIPK4
Clonality: Monoclonal
Clone: 2E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR