Anti-CCBE1 Rabbit Polyclonal Antibody

Supplier: Abnova
H00147372-D01
10581-342EA 376 USD
10581-342
Anti-CCBE1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CCBE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR