Anti-TMED6 (transmembrane emp24 Protein transport Domain Containing 6) Mouse Polyclonal Antibody

Supplier: Abnova
H00146456-B01P
10572-480EA 335 USD
10572-480
Anti-TMED6 (transmembrane emp24 Protein transport Domain Containing 6) Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TMED6 (transmembrane emp24 Protein transport Domain Containing 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR