Anti-RNF151 Mouse Monoclonal Antibody [clone: 4G8]

Supplier: Abnova
H00146310-M02
10599-588EA 335 USD
10599-588
Anti-RNF151 Mouse Monoclonal Antibody [clone: 4G8]
Antibodies
Type: Primary
Antigen: RNF151
Clonality: Monoclonal
Clone: 4G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR