Anti-CMTM4 Mouse Monoclonal Antibody [clone: 6G4]

Supplier: Abnova
H00146223-M01
10581-446EA 335 USD
10581-446
Anti-CMTM4 Mouse Monoclonal Antibody [clone: 6G4]
Antibodies
Type: Primary
Antigen: CMTM4
Clonality: Monoclonal
Clone: 6G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR