![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | TTBK2 |
Antigen Name | Tau Tubulin Kinase 2 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Mouse |
Western Blot | Yes |
Antigen Synonyms | TTBK2, TTBK, Tau-tubulin kinase 2, KIAA0847, SCA11 |
Gene ID | 146057 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |