Anti-SERPINA12 Mouse Monoclonal Antibody [clone: 3B3]

Supplier: Abnova
H00145264-M01
10600-978EA 335 USD
10600-978
Anti-SERPINA12 Mouse Monoclonal Antibody [clone: 3B3]
Antibodies
Type: Primary
Antigen: SERPINA12
Clonality: Monoclonal
Clone: 3B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR