![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | GSC |
Antigen Name | Goosecoid Homeobox |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Monoclonal |
Antibody Type | Primary |
Clone | 1C2 |
Host | Mouse |
Western Blot | Yes |
ELISA | Yes |
ImmunoChemistry | Yes |
Antigen Synonyms | SAMS |
Gene ID | 145258 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |