Anti-GSC Mouse Monoclonal Antibody [clone: 300]

Supplier: Abnova
H00145258-M13
10587-986EA 335 USD
10587-986
Anti-GSC Mouse Monoclonal Antibody [clone: 300]
Antibodies
Type: Primary
Antigen: GSC (goosecoid homeobox)
Clonality: Monoclonal
Clone: 300
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR