Anti-GSC Mouse Monoclonal Antibody [clone: 4B9]

Supplier: Abnova
H00145258-M10
10587-982EA 335 USD
10587-982
Anti-GSC Mouse Monoclonal Antibody [clone: 4B9]
Antibodies
Type: Primary
Antigen: GSC (goosecoid homeobox)
Clonality: Monoclonal
Clone: 4B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR