Anti-GSC Rabbit Polyclonal Antibody

Supplier: Abnova
H00145258-D01P
10587-808EA 376 USD
10587-808
Anti-GSC Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GSC (goosecoid homeobox)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR