Anti-LACC1 Mouse Monoclonal Antibody [clone: 2A1]

Supplier: Abnova
H00144811-M04
10586-372EA 335 USD
10586-372
Anti-LACC1 Mouse Monoclonal Antibody [clone: 2A1]
Antibodies
Type: Primary
Antigen: LACC1
Clonality: Monoclonal
Clone: 2A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR