Anti-GLIPR1L2 Mouse Polyclonal Antibody

Supplier: Abnova
H00144321-B01
10560-892EA 335 USD
10560-892
Anti-GLIPR1L2 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GLIPR1L2 (GLI pathogenesis-related 1 like 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR