![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | UBQLNL |
Antigen Name | Ubiquilin-like |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Mouse |
Western Blot | Yes |
Gene ID | 143630 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |