Anti-CACUL1 Mouse Polyclonal Antibody

Supplier: Abnova
H00143384-A01
10559-914EA 243 USD
10559-914
Anti-CACUL1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CACUL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR