Anti-HECTD2 Mouse Monoclonal Antibody [clone: 3D6]

Supplier: Abnova
H00143279-M01
10588-562EA 335 USD
10588-562
Anti-HECTD2 Mouse Monoclonal Antibody [clone: 3D6]
Antibodies
Type: Primary
Antigen: HECTD2
Clonality: Monoclonal
Clone: 3D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR