![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | MIB2 |
Antigen Name | Mindbomb E3 Ubiquitin Protein Ligase 2 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Monoclonal |
Antibody Type | Primary |
Clone | 3A7 |
Host | Mouse |
ELISA | Yes |
Antigen Synonyms | ZZANK1, ZZZ5 |
Gene ID | 142678 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |