Anti-SIRPA Mouse Monoclonal Antibody [clone: 1D10]

Supplier: Abnova
H00140885-M01
10579-142EA 335 USD
10579-142
Anti-SIRPA Mouse Monoclonal Antibody [clone: 1D10]
Antibodies
Type: Primary
Antigen: SIRPA
Clonality: Monoclonal
Clone: 1D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR