![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | SCP2D1 |
Antigen Name | SCP2 sterol-binding domain containing 1 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Mouse |
Western Blot | Yes |
Antigen Synonyms | HSD22, C20orf79, dJ1068E13.2 |
Gene ID | 140856 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |