Anti-ACTRT2 Mouse Monoclonal Antibody [clone: 20]

Supplier: Abnova
H00140625-M03
10580-998EA 335 USD
10580-998
Anti-ACTRT2 Mouse Monoclonal Antibody [clone: 20]
Antibodies
Type: Primary
Antigen: ACTRT2
Clonality: Monoclonal
Clone: 20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR