![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | ASB8 |
Antigen Name | Ankyrin Repeat And SOCS box Containing 8 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Mouse |
Western Blot | Yes |
Antigen Synonyms | PP14212 |
Gene ID | 140461 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |