![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | PASD1 |
Antigen Name | PAS Domain Containing 1 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Western Blot | Yes |
Antigen Synonyms | OXTES1, CT63 |
Gene ID | 139135 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |