![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | BEND2 |
Antigen Name | BEN Domain Containing 2 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Mouse |
Western Blot | Yes |
Antigen Synonyms | CXorf20 |
Gene ID | 139105 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |