Anti-UGT3A1 Mouse Polyclonal Antibody

H00133688-B01P
10573-448EA 316.92 USD
10573-448
Anti-UGT3A1 Mouse Polyclonal Antibody
Type: Primary
Antigen: UGT3A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR