Anti-SPATA4 Mouse Monoclonal Antibody [clone: 4A2]

Supplier: Abnova
H00132851-M03
10601-936EA 335 USD
10601-936
Anti-SPATA4 Mouse Monoclonal Antibody [clone: 4A2]
Antibodies
Type: Primary
Antigen: SPATA4
Clonality: Monoclonal
Clone: 4A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR