Anti-GRK7 Mouse Monoclonal Antibody [clone: 4C4]

Supplier: Abnova
H00131890-M02
10587-946EA 335 USD
10587-946
Anti-GRK7 Mouse Monoclonal Antibody [clone: 4C4]
Antibodies
Type: Primary
Antigen: GRK7 (G protein-coupled receptor kinase 7)
Clonality: Monoclonal
Clone: 4C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR