Anti-TPRA1 Mouse Monoclonal Antibody [clone: 6D7]

Supplier: Abnova
H00131601-M01
10604-516EA 335 USD
10604-516
Anti-TPRA1 Mouse Monoclonal Antibody [clone: 6D7]
Antibodies
Type: Primary
Antigen: TPRA1
Clonality: Monoclonal
Clone: 6D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR