Anti-DCBLD2 Mouse Monoclonal Antibody [clone: 3G10]

Supplier: Abnova
H00131566-M01
10583-580EA 335 USD
10583-580
Anti-DCBLD2 Mouse Monoclonal Antibody [clone: 3G10]
Antibodies
Type: Primary
Antigen: DCBLD2
Clonality: Monoclonal
Clone: 3G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR