Anti-FAM3D Mouse Polyclonal Antibody

Supplier: Abnova
H00131177-B01
10556-312EA 335 USD
10556-312
Anti-FAM3D Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FAM3D (family with sequence similarity 3, member D)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR