Anti-OSR1 Mouse Monoclonal Antibody [clone: 1C6]

Supplier: Abnova
H00130497-M31
10595-790EA 335 USD
10595-790
Anti-OSR1 Mouse Monoclonal Antibody [clone: 1C6]
Antibodies
Type: Primary
Antigen: OSR1
Clonality: Monoclonal
Clone: 1C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR