Anti-OSR1 Mouse Monoclonal Antibody [clone: 3F6]

H00130497-M25
10595-786EA 357.81 USD
10595-786
Anti-OSR1 Mouse Monoclonal Antibody [clone: 3F6]
Type: Primary
Antigen: OSR1
Clonality: Monoclonal
Clone: 3F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR