Anti-OSR1 Mouse Monoclonal Antibody [clone: 1G9]

Supplier: Abnova
H00130497-M10
10595-776EA 335 USD
10595-776
Anti-OSR1 Mouse Monoclonal Antibody [clone: 1G9]
Antibodies
Type: Primary
Antigen: OSR1
Clonality: Monoclonal
Clone: 1G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR