|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | OSR1 |
Antigen Name | Odd-skipped Related Transciption Factor 1 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Monoclonal |
Antibody Type | Primary |
Clone | 4B12 |
Host | Mouse |
ELISA | Yes |
Antigen Synonyms | Protein odd-skipped-related 1, OSR1, ODD |
Gene ID | 130497 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |