Anti-CNTNAP5 Mouse Polyclonal Antibody

Supplier: Abnova
H00129684-A01
10553-834EA 243 USD
10553-834
Anti-CNTNAP5 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CNTNAP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR