![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | IQGAP3 |
Antigen Name | IQ motif containing GTPase activating protein 3 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Western Blot | Yes |
Gene ID | 128239 |
Cross Adsorption | No |
Storage Temperature | Cool |
Shipping Temperature | Ice |