Anti-UHMK1 Mouse Monoclonal Antibody [clone: 1H5]

Supplier: Abnova
H00127933-M02
10591-100EA 335 USD
10591-100
Anti-UHMK1 Mouse Monoclonal Antibody [clone: 1H5]
Antibodies
Type: Primary
Antigen: UHMK1
Clonality: Monoclonal
Clone: 1H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR