Anti-GJB4 Mouse Monoclonal Antibody [clone: 1E3-1C12]

Supplier: Abnova
H00127534-M01
10587-402EA 335 USD
10587-402
Anti-GJB4 Mouse Monoclonal Antibody [clone: 1E3-1C12]
Antibodies
Type: Primary
Antigen: GJB4 (gap junction protein, beta 4, 30.3kDa)
Clonality: Monoclonal
Clone: 1E3-1C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR