Anti-GABPB2 Mouse Monoclonal Antibody [clone: 3A3-1F4]

Supplier: Abnova
H00126626-M01
10593-352EA 335 USD
10593-352
Anti-GABPB2 Mouse Monoclonal Antibody [clone: 3A3-1F4]
Antibodies
Type: Primary
Antigen: GABPB2 (GA binding protein transcription factor, beta subunit 2)
Clonality: Monoclonal
Clone: 3A3-1F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR