Anti-HSPB6 Mouse Monoclonal Antibody [clone: 6A4]

Supplier: Abnova
H00126393-M03
10589-492EA 335 USD
10589-492
Anti-HSPB6 Mouse Monoclonal Antibody [clone: 6A4]
Antibodies
Type: Primary
Antigen: HSPB6
Clonality: Monoclonal
Clone: 6A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR