Anti-ZNF577 Mouse Polyclonal Antibody

H00126231-B01P
10580-542EA 316.92 USD
10580-542
Anti-ZNF577 Mouse Polyclonal Antibody
Type: Primary
Antigen: ZNF577
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR